Methyl (2S,4S)-4-[2-nitro-4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051071-500MG 051071-2.5G
101894-176EA 364.62 USD
101894-176 101894-178
Methyl (2S,4S)-4-[2-nitro-4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-nitro-4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₄ClF₃N₂O₅
MDL Number: MFCD13561440
CAS Number: 1354486-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR