Methyl (2S,4S)-4-(2-chloro-4-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051069-500MG 051069-2.5G
101894-168EA 364.62 USD
101894-168 101894-170
Methyl (2S,4S)-4-(2-chloro-4-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-chloro-4-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄Cl₂N₂O₅
MDL Number: MFCD13561438
CAS Number: 1354488-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR