Methyl (2S,4S)-4-(2-methoxy-4-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051068-500MG 051068-2.5G
101894-164EA 364.62 USD
101894-164 101894-166
Methyl (2S,4S)-4-(2-methoxy-4-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-methoxy-4-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₄
MDL Number: MFCD13561437
CAS Number: 1354484-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR