Methyl (2S,4S)-4-{4-[(benzyloxy)carbonyl]phenoxy}-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051067-500MG 051067-2.5G
101894-160EA 364.62 USD
101894-160 101894-162
Methyl (2S,4S)-4-{4-[(benzyloxy)carbonyl]phenoxy}-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-{4-[(benzyloxy)carbonyl]phenoxy}-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₂ClNO₅
MDL Number: MFCD13561436
CAS Number: 1354487-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR