Methyl (2S,4S)-4-(2-iodophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051065-500MG 051065-2.5G
101894-152EA 364.62 USD
101894-152 101894-154
Methyl (2S,4S)-4-(2-iodophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-iodophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClINO₃
MDL Number: MFCD13561434
CAS Number: 1354485-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR