(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-iodophenoxy)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
051064-500MG
051064-2.5G
101894-148EA
461.54
USD
101894-148
101894-150
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-iodophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-iodophenoxy)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₆H₂₀INO₅ |
MDL Number:
MFCD13561433 CAS Number: 1354486-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...