Methyl (2S,4S)-4-(2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051063-500MG
051063-2.5G
101894-144EA
364.62
USD
101894-144
101894-146
Methyl (2S,4S)-4-(2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂O₅ |
MDL Number:
MFCD13561432 CAS Number: 1354487-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...