(2S,4S)-4-(2-Bromo-4-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051059-500MG 051059-2.5G
101894-128EA 364.62 USD
101894-128 101894-130
(2S,4S)-4-(2-Bromo-4-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Bromo-4-fluorophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉BrFNO₅
MDL Number: MFCD13561428
CAS Number: 1354484-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR