Methyl (2S,4S)-4-(4-methyl-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051057-500MG 051057-2.5G
101894-120EA 364.62 USD
101894-120 101894-122
Methyl (2S,4S)-4-(4-methyl-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-methyl-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₅
MDL Number: MFCD13561426
CAS Number: 1354488-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR