(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-methyl-2-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051056-500MG 051056-2.5G
101894-116EA 364.62 USD
101894-116 101894-118
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-methyl-2-nitrophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-methyl-2-nitrophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₇
MDL Number: MFCD13561425
CAS Number: 1354485-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR