Methyl (2S,4S)-4-(4-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051054-500MG 051054-2.5G
101894-108EA 364.62 USD
101894-108 101894-110
Methyl (2S,4S)-4-(4-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13561423
CAS Number: 1354488-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR