Methyl (2S,4S)-4-[4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051049-500MG 051049-2.5G
101894-088EA 364.62 USD
101894-088 101894-090
Methyl (2S,4S)-4-[4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[4-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₅ClF₃NO₃
MDL Number: MFCD13561418
CAS Number: 1354487-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR