Methyl (2S,4S)-4-[4-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051048-500MG 051048-2.5G
101894-084EA 364.62 USD
101894-084 101894-086
Methyl (2S,4S)-4-[4-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[4-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₅
MDL Number: MFCD13561417
CAS Number: 1354488-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR