(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051047-500MG 051047-2.5G
101894-080EA 364.62 USD
101894-080 101894-082
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₃NO₇
MDL Number: MFCD09862233
CAS Number: 1135201-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR