(2S,4S)-4-[4-(Acetylamino)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051044-500MG 051044-2.5G
101894-068EA 364.62 USD
101894-068 101894-070
(2S,4S)-4-[4-(Acetylamino)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-[4-(Acetylamino)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₆
MDL Number: MFCD13561414
CAS Number: 1354485-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR