Methyl (2S,4S)-4-(4-iodophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051043-500MG 051043-2.5G
101894-064EA 364.62 USD
101894-064 101894-066
Methyl (2S,4S)-4-(4-iodophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-iodophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClINO₃
MDL Number: MFCD13561413
CAS Number: 1354486-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR