Methyl (2S,4S)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051042-500MG 051042-2.5G
101894-060EA 364.62 USD
101894-060 101894-062
Methyl (2S,4S)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-chloro-3-methylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇Cl₂NO₃
MDL Number: MFCD13561412
CAS Number: 1354486-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR