Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051039-500MG
051039-2.5G
101894-048EA
364.62
USD
101894-048
101894-050
Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₃ |
MDL Number:
MFCD13561409 CAS Number: 1354487-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...