Methyl (2S,4S)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051038-500MG 051038-2.5G
101894-044EA 364.62 USD
101894-044 101894-046
Methyl (2S,4S)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[3-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13561408
CAS Number: 1354487-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR