Methyl (2S,4S)-4-[3-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051037-500MG
051037-2.5G
101894-040EA
364.62
USD
101894-040
101894-042
Methyl (2S,4S)-4-[3-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[3-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₀ClNO₅ |
MDL Number:
MFCD13561407 CAS Number: 1354484-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...