Methyl (2S,4S)-4-[3-(acetylamino)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051035-500MG 051035-2.5G
101894-032EA 364.62 USD
101894-032 101894-034
Methyl (2S,4S)-4-[3-(acetylamino)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[3-(acetylamino)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O₄
MDL Number: MFCD13561405
CAS Number: 1354484-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR