Methyl (2S,4S)-4-(5-isopropyl-2-methylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051033-500MG 051033-2.5G
101894-024EA 364.62 USD
101894-024 101894-026
Methyl (2S,4S)-4-(5-isopropyl-2-methylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(5-isopropyl-2-methylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13561403
CAS Number: 1354484-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR