Methyl (2S,4S)-4-[2-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051028-500MG 051028-2.5G
101894-004EA 364.62 USD
101894-004 101894-006
Methyl (2S,4S)-4-[2-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13561398
CAS Number: 1354488-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR