(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051027-500MG 051027-2.5G
101894-000EA 364.62 USD
101894-000 101894-002
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(tert-butyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₉NO₅
MDL Number: MFCD13561397
CAS Number: 1354486-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR