Methyl (2S,4S)-4-[2-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051026-500MG 051026-2.5G
101893-996EA 364.62 USD
101893-996 101893-998
Methyl (2S,4S)-4-[2-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-(trifluoromethyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₅ClF₃NO₃
MDL Number: MFCD13561396
CAS Number: 1354484-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR