Methyl (2S,4S)-4-(2-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051025-500MG
051025-2.5G
101893-992EA
364.62
USD
101893-992
101893-994
Methyl (2S,4S)-4-(2-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO₄ |
MDL Number:
MFCD13561395 CAS Number: 1354487-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...