(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4,5-trichlorophenoxy)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
051023-500MG
051023-2.5G
101893-984EA
364.62
USD
101893-984
101893-986
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4,5-trichlorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4,5-trichlorophenoxy)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₈Cl₃NO₅ |
MDL Number:
MFCD13561393 CAS Number: 1354486-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...