Methyl (2S,4S)-4-(2,4-dibromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051021-500MG 051021-2.5G
101893-976EA 364.62 USD
101893-976 101893-978
Methyl (2S,4S)-4-(2,4-dibromophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2,4-dibromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄Br₂ClNO₃
MDL Number: MFCD13561391
CAS Number: 1354487-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR