(2S,4S)-4-(2-Bromophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidine carboxylic acid

Supplier: Matrix Scientific

051018-500MG 051018-2.5G
101893-964EA 364.62 USD
101893-964 101893-966
(2S,4S)-4-(2-Bromophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidine carboxylic acid
(2S,4S)-4-(2-Bromophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidine carboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀BrNO₅
MDL Number: MFCD13561388
CAS Number: 1354485-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR