Methyl (2S,4S)-4-(4-chloro-3-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051017-500MG 051017-2.5G
101893-960EA 364.62 USD
101893-960 101893-962
Methyl (2S,4S)-4-(4-chloro-3-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-chloro-3-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉Cl₂NO₃
MDL Number: MFCD13561387
CAS Number: 1354488-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR