Methyl (2S,4S)-4-(4-acetylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051016-500MG 051016-2.5G
101893-956EA 364.62 USD
101893-956 101893-958
Methyl (2S,4S)-4-(4-acetylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-acetylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MDL Number: MFCD13561386
CAS Number: 1266111-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR