(2S,4S)-4-(4-Acetylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051015-500MG 051015-2.5G
101893-952EA 364.62 USD
101893-952 101893-954
(2S,4S)-4-(4-Acetylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Acetylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₃NO₆
MDL Number: MFCD13561385
CAS Number: 1266111-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR