Methyl (2S,4S)-4-(2-acetylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051014-500MG 051014-2.5G
101893-948EA 364.62 USD
101893-948 101893-950
Methyl (2S,4S)-4-(2-acetylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-acetylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄
MDL Number: MFCD13561384
CAS Number: 1266111-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR