Methyl (2S,4S)-4-[(5-chloro-8-quinolinyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051012-500MG 051012-2.5G
101893-940EA 364.62 USD
101893-940 101893-942
Methyl (2S,4S)-4-[(5-chloro-8-quinolinyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(5-chloro-8-quinolinyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₆Cl₂N₂O₃
MDL Number: MFCD13561382
CAS Number: 1354485-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR