Methyl (2S,4S)-4-(4-bromo-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051010-500MG 051010-2.5G
101893-932EA 364.62 USD
101893-932 101893-934
Methyl (2S,4S)-4-(4-bromo-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-bromo-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄BrClN₂O₅
MDL Number: MFCD13561380
CAS Number: 1354487-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR