(2S,4S)-4-(4-Bromo-2-nitrophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
051009-500MG
051009-2.5G
101893-928EA
364.62
USD
101893-928
101893-930
(2S,4S)-4-(4-Bromo-2-nitrophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Bromo-2-nitrophenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₉BrN₂O₇ |
MDL Number:
MFCD13561379 CAS Number: 1354485-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...