Methyl (2S,4S)-4-(4-chloro-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051008-500MG
051008-2.5G
101893-924EA
364.62
USD
101893-924
101893-926
Methyl (2S,4S)-4-(4-chloro-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-chloro-2-nitrophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₄Cl₂N₂O₅ |
MDL Number:
MFCD13561378 CAS Number: 1354487-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...