(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051005-500MG 051005-2.5G
101893-912EA 364.62 USD
101893-912 101893-914
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[4-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₅NO₇
MDL Number: MFCD13561375
CAS Number: 1354487-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR