(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

051003-500MG 051003-2.5G
101893-904EA 364.62 USD
101893-904 101893-906
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₃NO₇
MDL Number: MFCD13561373
CAS Number: 1354486-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR