Methyl (2S,4S)-4-[2-(tert-butyl)-4-methylphenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051000-500MG 051000-2.5G
101893-892EA 364.62 USD
101893-892 101893-894
Methyl (2S,4S)-4-[2-(tert-butyl)-4-methylphenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-(tert-butyl)-4-methylphenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₆ClNO₃
MDL Number: MFCD13561370
CAS Number: 1354487-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR