(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(tert-butyl)-4-methylphenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050999-500MG 050999-2.5G
101893-888EA 364.62 USD
101893-888 101893-890
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(tert-butyl)-4-methylphenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(tert-butyl)-4-methylphenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₁H₃₁NO₅
MDL Number: MFCD13561369
CAS Number: 1354486-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR