Methyl (2S,4S)-4-(4-allyl-2-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050998-500MG 050998-2.5G
101893-884EA 364.62 USD
101893-884 101893-886
Methyl (2S,4S)-4-(4-allyl-2-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-allyl-2-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₂ClNO₄
MDL Number: MFCD13561368
CAS Number: 1354488-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR