(2S,4S)-4-(4-Allyl-2-methoxyphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
050997-500MG
050997-2.5G
101893-880EA
364.62
USD
101893-880
101893-882
(2S,4S)-4-(4-Allyl-2-methoxyphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(4-Allyl-2-methoxyphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₂₀H₂₇NO₆ |
MDL Number:
MFCD13561367 CAS Number: 1354486-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...