Methyl (2S,4S)-4-[2-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050996-500MG 050996-2.5G
101893-876EA 364.62 USD
101893-876 101893-878
Methyl (2S,4S)-4-[2-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₅
MDL Number: MFCD13561366
CAS Number: 1354484-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR