(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050995-500MG 050995-2.5G
101893-872EA 364.62 USD
101893-872 101893-874
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[2-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₅NO₇
MDL Number: MFCD13561365
CAS Number: 1354486-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR