(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(5-chloro[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
050993-500MG
050993-2.5G
101893-864EA
364.62
USD
101893-864
101893-866
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(5-chloro[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(5-chloro[1,1'-biphenyl]-2-yl)oxy]-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₂₂H₂₄ClNO₅ |
MDL Number:
MFCD13561363 CAS Number: 1354486-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...