Methyl (2S,4S)-4-[2-bromo-4-(tert-pentyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
050982-500MG
050982-2.5G
101893-820EA
364.62
USD
101893-820
101893-822
Methyl (2S,4S)-4-[2-bromo-4-(tert-pentyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-bromo-4-(tert-pentyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₅BrClNO₃ MW: 406.76 g/mol |
MDL Number:
MFCD13561352 CAS Number: 1354484-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...