Methyl (2S,4S)-4-(4-chloro-2-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050980-500MG 050980-2.5G
101893-812EA 364.62 USD
101893-812 101893-814
Methyl (2S,4S)-4-(4-chloro-2-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-chloro-2-ethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉Cl₂NO₃
MDL Number: MFCD13561350
CAS Number: 1354488-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR