Methyl (2S,4S)-4-(2-chloro-4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050978-500MG 050978-2.5G
101893-804EA 364.62 USD
101893-804 101893-806
Methyl (2S,4S)-4-(2-chloro-4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-chloro-4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇Cl₂NO₄
MDL Number: MFCD13561348
CAS Number: 1354490-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR