(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-methoxyphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050977-500MG 050977-2.5G
101893-800EA 364.62 USD
101893-800 101893-802
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-methoxyphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-methoxyphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₂ClNO₆
MDL Number: MFCD13561347
CAS Number: 1354486-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR